missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113263
This item is not returnable.
View return policy
Description
RBM41 Polyclonal Antibody for Western Blot, IHC (P), ELISA
RBM41, known as RNA Binding Motif Protein 41, is identified as a novel component of the minor spliceosome, particularly interacting with its paralog, U11/U12-65K. Both RBM41 and U11/U12-65K share similar C-terminal RNA recognition motifs (RRMs), which are crucial for their role in spliceosomal activity. Distinctly, RBM41 has been associated with spliceosomal functions, although its precise cellular functions remain largely unexplored. Understanding the structural and functional parallels between RBM41 and U11/U12-65K aids in elucidating the mechanisms underlying minor spliceosome activity. The minor spliceosome itself is involved in the splicing of U12-type introns, thus playing a significant role in the regulation of gene expression. Additionally, RBM41's functional characterization in protein complexes and its involvement with RNA-binding activities enrich our understanding of gene regulation and cellular processes where the spliceosome plays a pivotal role. Research on RBM41 contributes to broader insights into RNA splicing and molecular genetics.
Specifications
| RBM41 | |
| Polyclonal | |
| Unconjugated | |
| RBM41 | |
| BB131279; D330023I21Rik; Rbm41; RNA binding motif protein 41; RNA-binding motif protein 41; RNA-binding protein 41 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55285 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96IZ5 | |
| RBM41 | |
| Recombinant Human RNA-binding protein 41 protein (1-413AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction