missing translation for 'onlineSavingsMsg'
Learn More

RBM33 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18410538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410538 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410538 Supplier Novus Biologicals Supplier No. NBP222280V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBM33 Polyclonal antibody specifically detects RBM33 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM33
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp434D1319, MGC20460, proline-rich protein 8, RNA binding motif protein 33, RNA-binding motif protein 33, RNA-binding protein 33
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 500 and 550 of human RNA Binding Motif Protein 33 using the numbering given in entry NP_444271.2 (GeneID 155435).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155435
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.