missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117673
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
RBM24 is preferentially expressed in muscle during differentiation in vitro and may regulate myogenic differentiation.
Specifications
| RBM24 | |
| Polyclonal | |
| Unconjugated | |
| rbm24a | |
| 6330546B05Rik; AI606861; dJ259A10.1; Rbm24; rbm24a; RNA binding motif protein 24; RNA binding motif protein 24a; RNA binding protein; rna-binding; RNA-binding motif protein 24; RNA-binding protein 24; RNA-binding protein SEB-4; RNA-binding region (RNP1, RRM) containing 6; RNA-binding region-containing protein 6; RNPC6; seb-4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 405801 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 4.34 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q76LC6 | |
| rbm24a | |
| Recombinant Zebrafish RNA-binding protein 24 protein (2-78aa). | |
| 100 μg | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction