missing translation for 'onlineSavingsMsg'
Learn More

RBM23 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18425588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425588 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425588 Supplier Novus Biologicals Supplier No. NBP281986AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBM23 Polyclonal antibody specifically detects RBM23 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM23
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CAPERbeta, FLJ10482, MGC4458, probable RNA-binding protein 23, RNA binding motif protein 23, RNA-binding motif protein 23, RNA-binding region (RNP1, RRM) containing 4, RNA-binding region-containing protein 4, RNPC4, Splicing factor SF2
Host Species Rabbit
Immunogen RBM23 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human RBM23. Amino Acid Squence: YRYGHSKSPHFREKSPVR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55147
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.