missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551618
This item is not returnable.
View return policy
Description
Immunogen sequence: NTYNRQNWED ADFPILCQTC LGENPYIRMT KEKYGKECKI CARPFTVFRW CPGVRMRFKK TEVCQTCSKL KNVCQTCLLD LEYGLPIQVR DAGLSFKDDM PKSDVNKEYY TQNMEREISN SDGTRPVGML GKATSTSDML LKL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes an RNA binding protein. The encoded protein may play a role in cell division and may be involved in pre-mRNA splicing. Related pseudogenes exist on chromosomes 6, 7, 9, 13, 16, 18, and X.
Specifications
| RBM22 | |
| Polyclonal | |
| Unconjugated | |
| RBM22 | |
| 199G4; 8430430L24Rik; Cwc2; fSAP47; functional spliceosome-associated protein 47; pre-mRNA-splicing factor RBM22; RBM22; RNA binding motif protein 22; RNA-binding motif protein 22; Unknown (protein for MGC:128706); ZC3H16; Zinc finger CCCH domain-containing protein 16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NW64 | |
| RBM22 | |
| Recombinant protein corresponding to Human RBM22. Recombinant protein control fragment (Product #RP-102115). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction