missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35096-100ul
This item is not returnable.
View return policy
Description
RBM22 Polyclonal antibody specifically detects RBM22 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RBM22 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Cwc2, FLJ10290, fSAP47, functional spliceosome-associated protein 47, pre-mRNA-splicing factor RBM22, RNA binding motif protein 22, ZC3H16RNA-binding motif protein 22, Zinc finger CCCH domain-containing protein 16 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human RBM22 (NP_060517.1).,, Sequence:, MATSLGSNTYNRQNWEDADFPILCQTCLGENPYIRMTKEKYGKECKICARPFTVFRWCPGVRMRFKKTEVCQTCSKLKNVCQTCLLDLEYGLPIQVRDAGLSFKDDMPKSDVNKEYYTQNMEREISNSDGTRPVGMLGKATSTSDMLLKLARTTPYYKRNRPHICSFWVKGECKRGEECPYRHEKPTDPDDPLADQNIKDRYYGINDPVADKLLKRASTMPRLDPPEDKT | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55696 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction