missing translation for 'onlineSavingsMsg'
Learn More

RBM22 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18414786 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414786 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414786 Supplier Novus Biologicals Supplier No. NBP222276AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBM22 Polyclonal antibody specifically detects RBM22 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM22
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Cwc2, FLJ10290, fSAP47, functional spliceosome-associated protein 47, pre-mRNA-splicing factor RBM22, RNA binding motif protein 22, ZC3H16RNA-binding motif protein 22, Zinc finger CCCH domain-containing protein 16
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 370 and 420 of human RNA Binding Motif Protein 22 using the numbering given in entry NP_060517.1 (GeneID 55696).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55696
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.