missing translation for 'onlineSavingsMsg'
Learn More

RBM19 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £366.00

Specifications

Antigen RBM19
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18358662
View Documents
Novus Biologicals
NBP3-05155-20UL
20 μL
£151.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18335455
View Documents
Novus Biologicals
NBP3-05155-100UL
100 μL
£366.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RBM19 Polyclonal specifically detects RBM19 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RBM19
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
9904
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human
KIAA0682DKFZp586F1023, NPO, probable RNA-binding protein 19, RNA binding motif protein 19, RNA-binding motif protein 19
Recombinant fusion protein containing a sequence corresponding to amino acids 759-960 of human RBM19 (NP_057280.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.