missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80576
This item is not returnable.
View return policy
Description
RBM17 Polyclonal specifically detects RBM17 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RBM17 | |
| Polyclonal | |
| Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_116294.1 | |
| RBM17 | |
| The immunogen this antibody was made to, maps to a region between residue 25 and 75 of human splicing factor (45kD) using the numbering given in entry NP_116294.1 (GeneID 84991). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| MGC14439, RNA binding motif protein 17, RNA-binding motif protein 17, SPF45DKFZp686F13131, splicing factor 45,45 kDa-splicing factor, splicing factor 45kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84991 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction