missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565572
This item is not returnable.
View return policy
Description
Immunogen sequence: RGARDRTPPLL YRDRDRDLYP DSDWVPPPPP VRERSTRTAA TSVPAYEPLD SLDRRRDGWS LDRDRGDRDL PSSRDQPRKR RLPEESGGRH LDRSPESDRP RKRHC Highest antigen sequence identity to the following orthologs - mouse 91%, rat 93%.
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.
Specifications
| RBM15 | |
| Polyclonal | |
| Unconjugated | |
| RBM15 | |
| C230088J01Rik; Kiaa4257; mKIAA1438; one twenty two protein; one twenty-two; one-twenty two protein 1; OTT; OTT1; putative RNA-binding protein 15; RBM15; RNA binding motif protein 15; RNA-binding motif protein 15; RNA-binding protein 15; SPEN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64783 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96T37 | |
| RBM15 | |
| Recombinant Human RBM15. Recombinant protein control fragment (Product #RP-104879). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction