missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-14796-100ul
This item is not returnable.
View return policy
Description
RBM14 Polyclonal antibody specifically detects RBM14 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RBM14 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:100-1:500 | |
| COAAMGC31756, Paraspeckle protein 2, PSP2DKFZp779J0927, RNA binding motif protein 14, RNA-binding motif protein 14, RNA-binding protein 14, RRM-containing coactivator activator/modulator, SIPMGC15912, Synaptotagmin-interacting protein, SYT-interacting protein, SYTIP1, TMEM137, transmembrane protein 137 | |
| Produced in rabbits immunized with E. coli-derived Human RBM14 fragment | |
| 100 μL | |
| Cell Cycle and Replication, DNA Repair | |
| 10432 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| Purified |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction