missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM12 Monoclonal Antibody (1D12)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520938
This item is not returnable.
View return policy
Description
Peptide Sequence: GPGPIHIGGP PGFASSSGKP GPTVIKVQNM PFTVSIDEIL DFFYGYQVIP GSVCLKYNEK GMPTGEAMVA FESRDEATAA VIDLNDRPIG SRKVKLVLG.
This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein.
Specifications
| RBM12 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NTZ6 | |
| RBM12 | |
| RBM12 (NP_006038.2, 834 a.a. ~ 932 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D12 | |
| Unconjugated | |
| RBM12 | |
| 5730420G12Rik; 9430070C08Rik; AI852903; Copine I; Copine-1; Cpne1; HRIHFB2091; KIAA0765; mKIAA0765; RBM12; RNA binding motif protein 12; RNA-binding motif protein 12; RNA-binding protein 12; SH3/WW domain anchor protein in the nucleus; SWAN | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10137 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction