missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540330
This item is not returnable.
View return policy
Description
Peptide sequence: MDYRSYPREY GSQEGKHDYD DSSEEQSAED SYEASPGSET QRRRRRRHRH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.
The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3' end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.
Specifications
| RBM10 | |
| Polyclonal | |
| Unconjugated | |
| RBM10 | |
| DXS8237E; E430039K10Rik; G patch domain-containing protein 9; GPATC9; GPATCH9; Kiaa0122; RBM10; RNA binding motif protein 10; rna-binding; RNA-binding motif protein 10; RNA-binding protein 10; RNA-binding protein S1-1; S1-1; S1-1 protein from liver; TARPS; ZRANB5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P98175 | |
| RBM10 | |
| Synthetic peptide directed towards the N-terminal of human RBM10 (aa 39-88). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction