missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBKS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580966
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RBKS/Ribokinase.
RBSK belongs to the pfkB family of carbohydrate kinases. It phosphorylates ribose to form ribose-5-phosphate in the presence of ATP and magnesium as a first step in ribose metabolism.
Specifications
| RBKS | |
| Polyclonal | |
| Unconjugated | |
| RBKS | |
| 5230400M11Rik; RBKS; RBSK; Ribokinase; RK | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 64080 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9H477 | |
| RBKS | |
| Recombinant Human RBKS/Ribokinase Protein (Met1-Phe322). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction