Learn More
Invitrogen™ RBG1L Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570753
Product Code. 15839163
  • £441.00 / 100µL
Description
This target displays homology in the following species: Dog: 86%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 100%.
RABGAP1L is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBG1L
Polyclonal
Unconjugated
Rabgap1l
5830411O09Rik; 8430421H08Rik; 9630005B12Rik; AW049894; expressed in hematopoietic cells, heart, liver (HLL); Hh1; Hhl; Kiaa0471; mKIAA0471; RAB GTPase activating protein 1 like; RAB GTPase activating protein 1-like; rab GTPase-activating protein 1-like; Rab GTPase-activating protein 1-like, isoform 10; RAB GTPase-activating protein 1-like-like protein; rab6 GTPase activating protein-like; RABGAP1L; RGD1304620; TBC1 domain family, member 18; TBC1D18
Rabbit
Affinity chromatography
RUO
9910
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
B7ZAP0, Q5R372
Rabgap1l
synthetic peptide directed towards the middle region of human RBG1L.