missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Mouse anti-Human, Mouse, Clone: CL11892, Novus Biologicals™
Mouse Monoclonal Antibody
£244.00 - £507.00
Specifications
| Antigen | RBFOX3/NeuN |
|---|---|
| Clone | CL11892 |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326415
|
Novus Biologicals
NBP3-15124-25UL |
25 μg |
£244.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18368291
|
Novus Biologicals
NBP3-15124-100UL |
100 μg |
£507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RBFOX3/NeuN | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3 | |
| This antibody was developed against a synthetic peptide. Actual immunogen sequence is proprietary information. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL11892 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| PBS (pH 7.2), 40% glycerol | |
| 146713 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title