missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBFOX2 Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143566
This item is not returnable.
View return policy
Description
RBFOX2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| RBFOX2 | |
| Polyclonal | |
| Unconjugated | |
| RBFOX2 | |
| dJ106I20.3; Fbm2; fox-1 homolog B; Fox-1 homolog Fxh; fox-1 homologue; FOX2; Fox-2; Fxh; Fyn-binding molecule 2; hexaribonucleotide-binding protein 2; HNRBP2; Hrnbp2; Rbfox2; Rbm9; repressor of tamoxifen transcriptional activity; RNA binding fox-1 homolog 2; RNA binding motif protein 9; RNA binding protein fox-1 homolog 2; RNA binding protein fox-1 homolog 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog 2; RNA-binding motif protein 9; RNA-binding protein 9; RTA; zgc:85694 | |
| Chicken | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| PBS with 5mM sodium azide | |
| A1A5R1, O43251, Q8BP71 | |
| RBFOX2 | |
| N-terminal 99 amino acids of human FOX2 expressed in and purified from E. coli. | |
| RUO | |
| 23543, 362950, 93686 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction