Learn More
Invitrogen™ RBFOX2 Polyclonal Antibody
Chicken Polyclonal Antibody
Brand:  Invitrogen™ PA5143532
 View more versions of this product
Product Code. 17845097
  • £1482.00 / 500µL
Description
RBFOX2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBFOX2
Polyclonal
Unconjugated
RBFOX2
dJ106I20.3; Fbm2; fox-1 homolog B; Fox-1 homolog Fxh; fox-1 homologue; FOX2; Fox-2; Fxh; Fyn-binding molecule 2; hexaribonucleotide-binding protein 2; HNRBP2; Hrnbp2; Rbfox2; Rbm9; repressor of tamoxifen transcriptional activity; RNA binding fox-1 homolog 2; RNA binding motif protein 9; RNA binding protein fox-1 homolog 2; RNA binding protein fox-1 homolog 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog 2; RNA-binding motif protein 9; RNA-binding protein 9; RTA; zgc:85694
Chicken
500 μL
Primary
Human, Mouse, Rat
Antibody
IgY
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Conc. Not Determined
PBS with 5mM sodium azide
A1A5R1, O43251, Q8BP71
RBFOX2
N-terminal 99 amino acids of human FOX2 expressed in and purified from E. coli.
RUO
23543, 362950, 93686
4°C