missing translation for 'onlineSavingsMsg'
Learn More

RBEL1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18408312 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408312 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408312 Supplier Novus Biologicals Supplier No. NBP212806AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBEL1 Polyclonal antibody specifically detects RBEL1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBEL1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias bA216L13.9, chromosome 9 open reading frame 86, FLJ10101, FLJ13045, KIAA1984, Parf, Partner of ARF, pp8875, putative GTP-binding protein Parf, Rab-like GTP-binding protein 1, Rab-like GTP-binding protein 1C, Rab-like protein 1, RBEL1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 375 and 425 of human Rab-like Protein 1 using the numbering given in entry NP_078994.3 (GeneID 55684).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55684
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.