missing translation for 'onlineSavingsMsg'
Learn More

RBEL1 Antibody (2E11-1F10), Novus Biologicals™

Product Code. 18349439 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349439 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349439 Supplier Novus Biologicals Supplier No. H00055684M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBEL1 Monoclonal antibody specifically detects RBEL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBEL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2E11-1F10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Sandwich ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH02945
Gene Alias bA216L13.9, chromosome 9 open reading frame 86, FLJ10101, FLJ13045, KIAA1984, Parf, Partner of ARF, pp8875, putative GTP-binding protein Parf, Rab-like GTP-binding protein 1, Rab-like GTP-binding protein 1C, Rab-like protein 1, RBEL1
Host Species Mouse
Immunogen C9orf86 (AAH02945, 1 a.a. ~ 477 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKIVIRGDRNTGKTALWHRLQGRPFVEEYIPTQEIQVTSIHWSYKTTDDIVKVEVWDVVDKGKCKKRGDGLKMENDPQEAESEMALDAEFLDVYKNCNGVVMMFDITKQWTFNYILRELPKVPTHVPVCVLGNYRDMGEHRVILPDDVRDFIDNLDRPPGSSYFRYAESSMKNSFGLKYLHKFFNIPFLQLQRETLLRQLETNQLDMDATLEELSVQQETEDQNYGIFLEMMEARSRGHASPLAANGQSPSPGSQSPVVPAGAVSTGSSSPGTPQPAPQLPLNAAPPSSVPPVPPSEALPPPACPSAPAPRRSIISRPPPWGWRLRGALGRRGQWPPWGGGRACHCLGRHLPLYHRLCRCPVAAVCASELEEAGHWWSPGWALQVLGLQAQCEPALQEGRGQLASARLGGHPGPLGAEPPVFLRDVTEAQEGPVRVCLQRLGRGRLAVGCALPRHLLALRAHLGPQHAYGSASGREP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55684
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.