missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBCK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £302.00
Specifications
| Antigen | RBCK1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-4ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18199074
|
Novus Biologicals
NBP2-27105SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18770274
|
Novus Biologicals
NBP2-27105 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBCK1 Polyclonal specifically detects RBCK1 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| RBCK1 | |
| Western Blot 2-4ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Primate | |
| Q9BYM8 | |
| 10616 | |
| A portion of amino acids 150-200 from human RBCk1 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS with 0.05% Sodium Azide | |
| C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4 | |
| RBCK1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, and chimpanzee, 94% in rhesus monkey, 89% in new world monkey, and macaque, 83% in cow, guinea pig, and mole rat, 78% in dog, elephant, and rabbit, 72% in panda, 67% in opossum, 61% in rat, and 55% homologous in mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title