missing translation for 'onlineSavingsMsg'
Learn More

RBC8, MedChemExpress

Product Code. 16418147
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418147

Brand: Medchem Express HY12873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RBC8 is a novel small molecule inhibitor of Ral GTPase; has IC50 of 3.5 μM in H2122 cell and 3.4 μM in H358 cell.

RBC8 is a novel small molecule inhibitor of Ral GTPase

Specifications

Chemical Name or Material RBC8
CAS 361185-42-4
Color White
Purity Grade Notes Research
Molecular Formula C25H20N4O3
Quantity 5 mg
Solubility Information DMSO : ≥ 40 mg/mL (94.24 mM)
SMILES N#CC(C1C2=CC(OC)=CC=C2OC)=C(N)OC3=C1C(C4=CC=C5C=CC=CC5=C4)=NN3
Molecular Weight (g/mol) 424.45
Formula Weight 424.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.