missing translation for 'onlineSavingsMsg'
Learn More

RBC8, MedChemExpress

Product Code. 16471607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471607 50 mg 50mg
16465428 10 mg 10mg
16426026 100 mg 100mg
16418147 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471607 Supplier Medchem Express Supplier No. HY12873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RBC8 is a novel small molecule inhibitor of Ral GTPase; has IC50 of 3.5 μM in H2122 cell and 3.4 μM in H358 cell.

RBC8 is a novel small molecule inhibitor of Ral GTPase

Specifications

Chemical Name or Material RBC8
CAS 361185-42-4
Color White
Purity Grade Notes Research
Molecular Formula C25H20N4O3
Quantity 50 mg
Solubility Information DMSO : ≥ 40 mg/mL (94.24 mM)
SMILES N#CC(C1C2=CC(OC)=CC=C2OC)=C(N)OC3=C1C(C4=CC=C5C=CC=CC5=C4)=NN3
Molecular Weight (g/mol) 424.45
Formula Weight 424.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.