missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBC Monoclonal Antibody (34-3C)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549100
This item is not returnable.
View return policy
Description
The monoclonal antibody only recognises antigenic determinants expressed on Mouse RBC and not on other species of RBC.
Red blood cells are the most common type of blood cell and are the vertebrate body's principal means of delivering oxygen from the lungs or gills to body tissues via the blood.
Specifications
| RBC | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and no preservative | |
| Mouse | |
| Protein G | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Functional Assay | |
| 34-3C | |
| Unconjugated | |
| Red Blood Cell | |
| RBC. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction