missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11882-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a cilia-associated protein belonging to the kinesin family. This protein plays a role in the sonic hedgehog (SHH) signaling pathway through the regulation of GLI transcription factors. It functions as a negative regulator of the SHH pathway by preventing inappropriate activation of GLI2 in the absence of ligand, and as a positive regulator by preventing the processing of GLI3 into its repressor form. Mutations in this gene have been associated with various ciliopathies.Specifications
| RBBP6 | |
| Polyclonal | |
| Unconjugated | |
| RBBP6 | |
| MY038, P2P R, P2PR, PACT, Protein P2P R, RBBP6, RBQ 1, RBQ1, SNAMA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19647, 5930 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97868, Q7Z6E9 | |
| RBBP6 | |
| RBBP6 Fusion Protein Ag2484 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction