Learn More
RBBP6 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Product Code. 18407947
Product Code. 18407947 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP149535MFV500
Rabbit Polyclonal Antibody
RBBP6 Polyclonal antibody specifically detects RBBP6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown