missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP6 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49535H
This item is not returnable.
View return policy
Description
RBBP6 Polyclonal specifically detects RBBP6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RBBP6 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q7Z6E9 | |
| RBBP6 | |
| A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 1600-1650) [UniProt Q7Z6E9] | |
| 0.1 mL | |
| Tumor Suppressors | |
| 5930 | |
| Human, Mouse | |
| IgG |
| KnockDown, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS with No Preservative | |
| E3 ubiquitin-protein ligase RBBP6, EC 6.3.2.-, MY038, P2PR, P2P-R, p53-associated cellular protein of testis, PACTDKFZp761B2423, Proliferation potential-related protein, Protein P2P-R, RB-binding Q-protein 1, RBQ1, RBQ-1, retinoblastoma binding protein 6, Retinoblastoma-binding protein 6DKFZp686P0638, Retinoblastoma-binding Q protein 1, SNAMA | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody may detect isoform 1 (201.6 kDa), isoform 2 (197.3 kDa) and isoform 4 (106.0 kDa) of human RBBP6. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction