missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP6 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49535AF350
This item is not returnable.
View return policy
Description
RBBP6 Polyclonal specifically detects RBBP6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RBBP6 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| E3 ubiquitin-protein ligase RBBP6, EC 6.3.2.-, MY038, P2PR, P2P-R, p53-associated cellular protein of testis, PACTDKFZp761B2423, Proliferation potential-related protein, Protein P2P-R, RB-binding Q-protein 1, RBQ1, RBQ-1, retinoblastoma binding protein 6, Retinoblastoma-binding protein 6DKFZp686P0638, Retinoblastoma-binding Q protein 1, SNAMA | |
| Rabbit | |
| 0.1 mL | |
| 5930 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RBBP6 | |
| A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 1600-1650) [UniProt Q7Z6E9] | |
| Primary | |
| This antibody may detect isoform 1 (201.6 kDa), isoform 2 (197.3 kDa) and isoform 4 (106.0 kDa) of human RBBP6. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction