missing translation for 'onlineSavingsMsg'
Learn More

RbBP5 Antibody, Novus Biologicals™

Product Code. 18349459 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349459 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349459 Supplier Novus Biologicals Supplier No. H00005929B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RbBP5 Polyclonal antibody specifically detects RbBP5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RbBP5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005048
Gene Alias RBQ3RBBP-5, retinoblastoma binding protein 5, Retinoblastoma-binding protein RBQ-3, Set1c WD40 repeat protein, homolog, SWD1
Host Species Mouse
Immunogen RBBP5 (NP_005048, 1 a.a. - 538 a.a.) full-length human protein. MNLELLESFGQNYPEEADGTLDCISMALTCTFNRWGTLLAVGCNDGRIVIWDFLTRGIAKIISAHIHPVCSLCWSRDGHKLVSASTDNIVSQWDVLSGDCDQRFRFPSPILKVQYHPRDQNKVLVCPMKSAPVMLTLSDSKHVVLPVDDDSDLNVVASFDRRGEYIYTGNAKGKILVLKTDSQDLVASFRVTTGTSNTTAIKSIEFARKGSCFLINTADRIIRVYDGREILTCGRDGEPEPMQKLQDLVNRTPWKKCCFSGDGEYIVAGSARQHALYIWEKSIGNLVKILHGTRGELLLDVAWHPVRPIIASISSGVVSIWAQNQVENWSAFAPDFKELDENVEYEERESEFDIEDEDKSEPEQTGADAAEDEEVDVTSVDPIAAFCSSDEELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKGKEKDSPFKPKLYKGDRGLPLEGSAKGKVQAELSQPLTAGGAISELL
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 5929
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.