missing translation for 'onlineSavingsMsg'
Learn More

RbBP5 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18431794 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431794 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431794 Supplier Novus Biologicals Supplier No. NB600252FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RbBP5 Polyclonal antibody specifically detects RbBP5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RbBP5
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias RBQ3RBBP-5, retinoblastoma binding protein 5, Retinoblastoma-binding protein RBQ-3, Set1c WD40 repeat protein, homolog, SWD1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and the C-terminus (residue 538) of human retinoblastoma binding protein 5 using the numbering given in entry NP_005048.2 (GeneID 5929).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 5929
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.