missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP4/RbAp48 Antibody (3E9), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £431.00
Specifications
| Antigen | RBBP4/RbAp48 |
|---|---|
| Clone | 3E9 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030563
|
Novus Biologicals
NBP3-26350-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032741
|
Novus Biologicals
NBP3-26350-100ul |
100 μL |
£431.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBBP4/RbAp48 Monoclonal antibody specifically detects RBBP4/RbAp48 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| RBBP4/RbAp48 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48 | |
| A synthesized peptide derived from Human RBBP4/RbAp48 [UniProt Q09028] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3E9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5928 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title