missing translation for 'onlineSavingsMsg'
Learn More

RBBP4/RbAp48 Antibody (2D7), Novus Biologicals™

Product Code. 18323859 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323859 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323859 Supplier Novus Biologicals Supplier No. H00005928M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBBP4/RbAp48 Monoclonal antibody specifically detects RBBP4/RbAp48 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBBP4/RbAp48
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH53904
Gene Alias CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48
Host Species Mouse
Immunogen RBBP4 (AAH53904, 1 a.a. ~ 425 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MADKEAAFDDAVEERVINEEYKIWKKNTPFLYDLVMTHALEWPSLTAQWLPDVTRPEGKDFSIHRLVLGTHTSDEQNHLVIASVQLPNDDAQFDASHYDSEKGEFGGFGSVSGKIEIEIKINHEGEVNRARYMPQNPCIIATKTPSSDVLVFDYTKHPSKPDPSGECNPDLRLRGHQKEGYGLSWNPNLSGHLLSASDDHTICLWDISAVPKEGKVVDAKTIFTGHTAVVEDVSWHLLHESLFGSVADDQKLMIWDTRSNNTSKPSHSVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHSFESHKDEIFQVQWSPHNETILASSGTDRRLNVWDLSKIGEEQSPEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQVWQMAENIYNDEDPEGSVDPEGQGS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5928
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.