missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP4/RbAp48 Antibody (15G12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41202
This item is not returnable.
View return policy
Description
RBBP4/RbAp48 Monoclonal specifically detects RBBP4/RbAp48 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| RBBP4/RbAp48 | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 0.2-1 ug/ml, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| RBBP4 | |
| The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli. | |
| 100 μL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors | |
| 5928 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 15G12 | |
| Unconjugated | |
| CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| This antibody will recognize human RbAp48 + RbAp46. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction