missing translation for 'onlineSavingsMsg'
Learn More

RBBP4/RbAp48 Antibody (13D10), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications

Brand:  Novus Biologicals NBP1-41201

Product Code. 18253281

  • £449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RBBP4/RbAp48 Monoclonal specifically detects RBBP4/RbAp48 in Human, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RBBP4/RbAp48
Monoclonal
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:500, Chromatin Immunoprecipitation (ChIP)
RBBP4
The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli.
Immunogen affinity purified
RUO
Primary
This antibody will recognize human RbAp48.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
13D10
Unconjugated
CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48
Mouse
48 kDa
100 μL
Cancer, Cell Cycle and Replication, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors
5928
Human, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only