Learn More
RBBP4 Mouse anti-Human, Clone: 11G10, Abnova™
Mouse monoclonal antibody raised against human RBBP4.
Brand: Abnova MAB12526.50uL
Description
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. It is present in protein complexes involved in histone acetylation and chromatin assembly. It is part of the Mi-2 complex which has been implicated in chromatin remodeling and transcriptional repression associated with histone deacetylation. This encoded protein is also part of co-repressor complexes, which is an integral component of transcriptional silencing. It is found among several cellular proteins that bind directly to retinoblastoma protein to regulate cell proliferation. This protein also seems to be involved in transcriptional repression of E2F-responsive genes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| RBBP4 | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user. | |
| NURF55/RBAP48 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5928 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 11G10 | |
| Mouse monoclonal antibody raised against human RBBP4. | |
| In PBS. | |
| RBBP4 | |
| Recombinant protein corresponding to amino acids 1-425 of human RBBP4. | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.