missing translation for 'onlineSavingsMsg'
Learn More

RB670 Mouse Anti-Human CD44, BD OptiBuild™

Product Code. 31792181 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31792181 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31792181 Supplier BD Biosciences Supplier No. 771092
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD44 Monoclonal Antibody detects CD44 in Human Samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone 515
Concentration 0.2 mg/mL
Conjugate RB670
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Pgp-1; CSPG8; ECMR-III; Epican; H-CAM; HCELL; Hermes; HUTCH-1
Host Species Mouse
Immunogen Human EBV-transformed Arent B Lymphoblastoid Cell Line
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.