missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1CC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17250-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1CC1. the calculated molecular weight of RB1CC1 is 180 kDa, but the modified RB1CC1 is about 200 kDa.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
| RB1CC1 | |
| Polyclonal | |
| Unconjugated | |
| RB1CC1 | |
| CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12421, 312927, 9821 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TDY2, Q9ESK9 | |
| Rb1cc1 | |
| RB1CC1 Fusion Protein Ag10508 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction