missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1CC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93785-0.02ml
This item is not returnable.
View return policy
Description
RB1CC1 Polyclonal antibody specifically detects RB1CC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RB1CC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| CC1, DRAGOU14, FIP200, KIAA0203200 kDa FAK family kinase-interacting protein, RB1-inducible coiled-coil 1, RB1-inducible coiled-coil protein 1, RBICC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1280-1400 of human RB1CC1 (NP_055596.3). TRGDSSSLVAELQEKLQEEKAKFLEQLEEQEKRKNEEMQNVRTSLIAEQQTNFNTVLTREKMRKENIINDLSDKLKSTMQQQERDKDLIESLSEDRARLLEEKKKLEEEVSKLRSSSFVPS | |
| 0.02 mL | |
| Tumor Suppressors | |
| 9821 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction