missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM15B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557749
This item is not returnable.
View return policy
Description
Immunogen sequence: GKKARDSERN HRTTEAEPKP LEEPKHETKK LKNLSEYAQT LQLGWNGLLV LKNSCFPTSM HILEGDQGVI SSLLKDHTSG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
May function in the regulation of alternative or illicit splicing.
Specifications
| RBM15B | |
| Polyclonal | |
| Unconjugated | |
| RBM15B | |
| 1810017N16Rik; chromosome 3p21.1 gene sequence; HsOTT3; HUMAGCGB; huOTT3; one twenty two protein 3; one-twenty two protein 3; OTT3; putative RNA-binding protein 15B; Rbm15b; RGD1305937; RNA binding motif protein 15 pseudogene; RNA binding motif protein 15B; rna-binding; RNA-binding motif protein 15B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NDT2 | |
| RBM15B | |
| Recombinant protein corresponding to Human RB15B. Recombinant protein control fragment (Product #RP-96501). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction