missing translation for 'onlineSavingsMsg'
Learn More

RB1 Rabbit anti-Human, Mouse, Clone: RB1/2313R, Novus Biologicals™

Product Code. 18374032 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374032 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374032 Supplier Novus Biologicals Supplier No. NBP30719620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RB1 Monoclonal specifically detects RB1 in Human, Mouse samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RB1/2313R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Host Species Rabbit
Immunogen Recombinant human full-length RB1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5925
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.