Learn More
RB1, p Ser811 Antibody (S01-2G6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19969-50ul
 View more versions of this product
Product Code. 18657004
  • £328.00 / 50µL
Description
RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RB1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S01-2G6
Unconjugated
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
A synthetic phosphopeptide corresponding to residues surrounding Ser811 of human RB1 (Uniprot # P06400)
50 μg
Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
5925
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.