missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 [p Ser780] Antibody (JE44-61), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32794
This item is not returnable.
View return policy
Description
RB1 Monoclonal antibody specifically detects RB1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RB1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| Synthetic phosphopeptide corresponding to resudues surrounding Ser780 of human RB1 protein. (Uniprot: P06400) | |
| 100 μL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 5925 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE44-61 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction