missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Antibody (S09-8G7), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £501.00
Specifications
| Antigen | RB1 |
|---|---|
| Clone | S09-8G7 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630705
|
Novus Biologicals
NBP3-19936-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608704
|
Novus Biologicals
NBP3-19936-100ul |
100 μg |
£501.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RB1 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| Recombinant protein of mouse RB1 (Uniprot # P13405) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S09-8G7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5925 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title