missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rb Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5110276
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: RILVSIGESF GTSEKFQKIN QMVCNSDRVL KRSAEGSNPP KPLKKLRFDI EGSDEADGSK HLPGESKFQQ KLAEMTSTRT RMQKQKMNDS MDTSNKEEK
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.
Specifications
| Rb | |
| Polyclonal | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06400, P13405 | |
| RB1 | |
| A synthetic peptide corresponding to a sequence within amino acids 830 to the C-terminus of human Rb (NP_000312.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| RB1 | |
| Cleaved Rb; delta RB-p70; DRb-p70; OSRC; p104; P105RB; p105-Rb; Phospho-Retinoblastoma; pp105; pp110; PPP1R130; pRb; Rb; RB1; Rb-1; Rb-p70; Retin; Retinoblastoma; Retinoblastoma-associated protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19645, 5925 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction