missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rb Monoclonal Antibody (1F8 (Rb1)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511387
This item is not returnable.
View return policy
Description
MA5-11387 targets Retinoblastoma in FACS, ICC/IF, IHC (P), IP, and WB applications and shows reactivity with human and mouse samples. Does not react with rat samples. The MA5-11387 immunogen is recombinant human Rb protein.
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.
Specifications
| Rb | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P06400, P13405 | |
| RB1 | |
| Recombinant human Rb protein | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1F8 (Rb1) | |
| Unconjugated | |
| RB1 | |
| Cleaved Rb; delta RB-p70; DRb-p70; exon 17 tumor GOS561 substitution mutation causes premature stop; GOS563 exon 17 substitution mutation causes premature stop; OSRC; p104; p104 chicken Rb; P105RB; p105-Rb; Phospho-Retinoblastoma; pp105; pp110; PPP1R130; pRb; prepro-retinoblastoma-associated protein; protein phosphatase 1, regulatory subunit 130; rb; RB transcriptional corepressor 1; RB1; Rb-1; Rb-p70; Retin; Retinoblastoma; retinoblastoma 1; retinoblastoma 1 (including osteosarcoma); retinoblastoma 1 protein; retinoblastoma gene; retinoblastoma isoform A; retinoblastoma protein; retinoblastoma suspectibility protein; retinoblastoma tumor suppressor; retinoblastoma-associated protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 19645, 5925 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction