missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB associated KRAB repressor Antibody (6F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057786-M01
This item is not returnable.
View return policy
Description
RB associated KRAB repressor Monoclonal antibody specifically detects RB associated KRAB repressor in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| RB associated KRAB repressor | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| hRBaK, RB-associated KRAB repressor, RB-associated KRAB zinc finger, RB-associated KRAB zinc finger protein, ZNF769Zinc finger protein 769 | |
| RBAK (NP_066986, 51 a.a. ∽ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TTKPNVIIKLEQGEEPWIMGGEFPCQHSPEAWRVDDLIERIQENEDKHSRQAACINSKTLTEEKENTFSQIYMETSLVPSSIIAHNCVSCGKNLESISQL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 6F9 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_066986 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 57786 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction