Learn More
EduLab™ Ray Box
Product Code. 13152920
Product Code. 13152920 Supplier EduLab™ Supplier No. EDU1207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for a wide range of optics experiments.
  • Designed with two slots to hold blanking plates, seven grooves for lenses
  • Supplied with 12V, 20 - 24W, bulb, holder, slit plates, blanking plates and instructions
Specifications
Includes 12V, 20 - 24W, bulb, holder, slit plates, blanking plates and instructions
Product Type Ray Box