missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAX Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56467DL594
This item is not returnable.
View return policy
Description
RAX Polyclonal specifically detects RAX in Human, Canine, Primate samples. It is validated for Western Blot.
Specifications
| RAX | |
| Polyclonal | |
| Western Blot | |
| MCOP3, retina and anterior neural fold homeobox, retinal homeobox protein Rx, RXRetina and anterior neural fold homeobox protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| The amino acid sequence used as immunogen has 100% identity in human, dog, and monkey. | |
| Store at 4°C in the dark. |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAX | |
| A portion of amino acids 300-350 of human Rax was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction