missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAVER2 Recombinant Rabbit Monoclonal Antibody (K01_1X64)
Description
Immunogen sequence: PNQHIAGQAG PGHSNTQEKQ PATVGMAEGN FSGSQPYLQS FPNLAAGSLL VGHHKQQQSQ PKGTEISSGA ASKNQTSLLG EPPKEIRLSK NPYLNLASVL PSVCLSSPAS KTTLHKTGIA SSILDAISQG SESQHALEKC IAYSPPFGDY AQVSSLRNEK RGSSYLISAP EGGSVECVDQ HSQGTGAYYM ETYLKKKR Highest antigen sequence identity to the following orthologs: Mouse - 75%, Pig - 86%, Cynomolgus monkey - 98%.
RAVER2 may bind single-stranded nucleic acids.
Specifications
Specifications
| Antigen | RAVER2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | K01_1X64 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | RAVER2 |
| Gene Accession No. | Q9HCJ3 |
| Gene Alias | A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?