missing translation for 'onlineSavingsMsg'
Learn More

RAVER1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18429396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429396 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429396 Supplier Novus Biologicals Supplier No. NBP222291AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAVER1 Polyclonal antibody specifically detects RAVER1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAVER1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA1978Protein raver-1, ribonucleoprotein PTB-binding 1, ribonucleoprotein, PTB-binding 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 556 and 606 of human Ribonucleoprotein, PTB-Binding using the numbering given in entry Q8IY67.1 (GeneID 125950).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125950
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.